Merck
  • Soluble platelet endothelial cell adhesion molecule 1 (sPECAM-1) improves diagnostic accuracy of D-Dimer in patients with suspected deep vein thrombosis (DVT).
Soluble platelet endothelial cell adhesion molecule 1 (sPECAM-1) improves diagnostic accuracy of D-Dimer in patients with suspected deep vein thrombosis (DVT).
Journal of thrombosis and thrombolysis (2020-03-28)
Joerg Kellermair, Alexander Fellner, Alexander Bittinger, Stephanie Schneider, Kaveh Akbari, Juergen Kammler, Thomas Lambert, Clemens Steinwender
ABSTRACT
D-Dimer has a high sensitivity but a low specificity for the diagnosis of deep vein thrombosis (DVT) which limits its implementation as a general screening parameter. There is a demand for additional biomarkers to improve its diagnostic accuracy. Soluble platelet endothelial cell adhesion molecule 1 (sPECAM-1) is generated at the site of venous thrombosis, thus, represents a promising biomarker. Patients with clinically suspected DVT (N = 159) were prospectively recruited and underwent manual compression ultrasonography (CCUS) to confirm or exclude DVT. The diagnostic value of D-Dimer, sPECAM-1 and the combination of both was assessed. sPECAM-1 levels were significantly higher in patients with DVT (N = 44) compared to patients without DVT (N = 115) (85.9 [76.1/98.0] ng/mL versus 68.0 [50.1/86.0] ng/mL; p < 0.001) with a diagnostic sensitivity of 100% and a specificity of 28.7% at the cut point > 50.2 ng/mL. sPECAM-1 improved the diagnostic accuracy of D-Dimer: the combination of both biomarkers yielded a ROC-AUC of 0.925 compared to 0.905 for D-Dimer alone and 0.721 for sPECAM-1 alone with a reduction of false-positive D-Dimer cases 72- > 43 (Δ =  - 31.9%). The discrimination mainly occurred in a subgroup of patients characterized by an inflammatory background (defined by c-reactive protein level > 1 mg/mL). sPECAM-1 represents a novel diagnostic biomarker for venous thrombosis. It does not qualify as a diagnostic biomarker alone but improves the diagnostic accuracy of D-Dimer in patients with suspected DVT.