Merck
  • A Longitudinal Study of the Antioxidant Barrier and Oxidative Stress in Morbidly Obese Patients after Bariatric Surgery. Does the Metabolic Syndrome Affect the Redox Homeostasis of Obese People?
A Longitudinal Study of the Antioxidant Barrier and Oxidative Stress in Morbidly Obese Patients after Bariatric Surgery. Does the Metabolic Syndrome Affect the Redox Homeostasis of Obese People?
Journal of clinical medicine (2020-04-05)
Barbara Choromańska, Piotr Myśliwiec, Magdalena Łuba, Piotr Wojskowicz, Jacek Dadan, Hanna Myśliwiec, Katarzyna Choromańska, Anna Zalewska, Mateusz Maciejczyk
ABSTRACT
This is the first study to evaluate both the antioxidant barrier, glutathione metabolism, and oxidative damage to proteins and lipids in morbidly obese patients undergoing bariatric treatment. The study included 65 patients with class 3 obesity divided into two subgroups: morbidly obese patients without metabolic syndrome (OB) and obese patients with metabolic syndrome (OB + MS). Blood samples were collected before surgery as well as one, three, six, and twelve months after the bariatric treatment. Superoxide dismutase and reduced glutathione (GSH) were significantly decreased, whereas glutathione reductase and uric acid were enhanced in morbidly obese patients before bariatric surgery as compared to lean control. Moreover, in the OB group, we observed the increase of superoxide dismutase (SOD) and the decrease of uric acid (UA) after the bariatric treatment; however, these changes were not observed in the OB + MS group. The oxidative damage to proteins (advanced glycation end products, AGE; advanced oxidation protein products, AOPP) and lipids (8-isoprostanes, 8-isop; 4-hydroxynoneal) was higher in OB as well as OB + MS patients. We noticed that AGE and AOPP levels diminished after the bariatric treatment, whereas redox status (ratio of GSH to oxidized glutathione) was still reduced in the OB + MS group. Summarizing, morbid obesity is associated with disturbances in the antioxidant barrier and enhanced oxidative damage to proteins and lipids. Although bariatric surgery improves redox homeostasis in obese patients, those with metabolic syndrome show a continuous decrease in the antioxidant status. In patients undergoing bariatric treatment, antioxidant supplementation may be considered.