Merck
  • A Humanized Lym-1 CAR with Novel DAP10/DAP12 Signaling Domains Demonstrates Reduced Tonic Signaling and Increased Antitumor Activity in B-Cell Lymphoma Models.
A Humanized Lym-1 CAR with Novel DAP10/DAP12 Signaling Domains Demonstrates Reduced Tonic Signaling and Increased Antitumor Activity in B-Cell Lymphoma Models.
Clinical cancer research : an official journal of the American Association for Cancer Research (2020-04-11)
Long Zheng, Luqing Ren, Aida Kouhi, Leslie A Khawli, Peisheng Hu, Harvey R Kaslow, Alan L Epstein
ABSTRACT
The murine Lym-1 mAb targets a discontinuous epitope (Lym-1 epitope) on several subtypes of HLA-DR, which is upregulated in a majority of human B-cell lymphomas and leukemias. Unlike CD19, the Lym-1 epitope does not downregulate upon crosslinking, which may provide an advantage as a target for CAR T-cell therapy. Lym-1 CAR T cells with a conventional 4-1BB and CD3ζ (BB3z) signaling domain exhibited impaired ex vivo expansion. This study aimed to identify the underlying mechanisms and develop strategies to overcome this effect. A functional humanized Lym-1 antibody (huLym-1-B) was identified and its scFv form was used for CAR design. To overcome observed impaired expansion in vitro, a huLym-1-B CAR using DAP10 and DAP12 (DAP) signaling domains was evaluated for ex vivo expansion and in vivo function. Impaired expansion in huLym-1-B-BB3z CAR T cells was shown to be due to ligand-dependent suboptimal CAR signaling caused by interaction of the CAR binding domain and the surface of human T cells. Using the novel DAP signaling domain construct, the effects of suboptimal CAR signaling were overcome to produce huLym-1-B CAR T cells with improved expansion ex vivo and function in vivo. In addition, the Lym-1 epitope does not significantly downregulate in response to huLym-1-B-DAP CAR T cells both ex vivo and in vivo. DAP intracellular domains can serve as signaling motifs for CAR, and this new construct enables nonimpaired production of huLym-1-B CAR T cells with potent in vivo antitumor efficacy.
Product Description
Millipore
D-(+)-Trehalose dihydrate, suitable for microbiology, ≥99.0%, composed of two α-glucose units that is used as a protectant, stabilizer and to support proper folding
Supelco
Calcium Standard for ICP, TraceCERT®, 1 g/L Ca in nitric acid (nominal concentration)
Sigma-Aldrich
Click′s Medium, With sodium bicarbonate, without mercaptoethanol and L-glutamine, liquid, sterile-filtered, suitable for cell culture