Merck
  • Integral approach to biomacromolecular structure by analytical-ultracentrifugation and small-angle scattering.
Integral approach to biomacromolecular structure by analytical-ultracentrifugation and small-angle scattering.
Communications biology (2020-06-10)
Ken Morishima, Aya Okuda, Rintaro Inoue, Nobuhiro Sato, Yosuke Miyamoto, Reiko Urade, Maho Yagi-Utsumi, Koichi Kato, Rina Hirano, Tomoya Kujirai, Hitoshi Kurumizaka, Masaaki Sugiyama
ABSTRACT
Currently, a sample for small-angle scattering (SAS) is usually highly purified and looks monodispersed: The Guinier plot of its SAS intensity shows a fine straight line. However, it could include the slight aggregates which make the experimental SAS profile different from the monodispersed one. A concerted method with analytical-ultracentrifugation (AUC) and SAS, named as AUC-SAS, offers the precise scattering intensity of a concerned biomacromolecule in solution even with aggregates as well that of a complex under an association-dissociation equilibrium. AUC-SAS overcomes an aggregation problem which has been an obstacle for SAS analysis and, furthermore, has a potential to lead to a structural analysis for a general multi-component system.
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Apoferritin from equine spleen, 0.2 μm filtered