Merck
  • Celecoxib substituted biotinylated poly(amidoamine) G3 dendrimer as potential treatment for temozolomide resistant glioma therapy and anti-nematode agent.
Celecoxib substituted biotinylated poly(amidoamine) G3 dendrimer as potential treatment for temozolomide resistant glioma therapy and anti-nematode agent.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2020-07-03)
Łukasz Uram, Joanna Markowicz, Maria Misiorek, Aleksandra Filipowicz-Rachwał, Stanisław Wołowiec, Elżbieta Wałajtys-Rode
ABSTRACT
Glioblastoma multiforme (GBM) is a one of the most widely diagnosed and difficult to treat type of central nervous system tumors. Resection combined with radiotherapy and temozolomide (TMZ) chemotherapy prolongs patients' survival only for 12 - 15 months after diagnosis. Moreover, many patients develop TMZ resistance, thus important is search for a new therapy regimes including targeted drug delivery. Most types of GBM reveal increased expression of cyclooxygenase-2 (COX-2) and production of prostaglandin E2 (PGE2), that are considered as valuable therapeutic target. In these studies, the anti-tumor properties of the selective COX-2 inhibitor celecoxib (CXB) and biotinylated third generation of the poly(amidoamine) dendrimer substituted with 31 CXB residues (G3BC31) on TMZ -resistant U-118 MG glioma cell line were examined and compared with the effect of TMZ alone including viability, proliferation, migration and apoptosis, as well as the cellular expression of COX-2, ATP level, and PGE2 production. Confocal microscopy analysis with the fluorescently labeled G3BC31 analogue has shown that the compound was effectively accumulated in U-118 MG cells in time-dependent manner and its localization was confirmed in lysosomes but not nuclei. G3BC31 reveal much higher cytotoxicity for U-118 MG cells at relatively low concentrations in the range of 2-4 µM with compared to CBX alone, active at 50-100 µM. This was due to induction of apoptosis and inhibition of proliferation and migration. Observed effects were concomitant with reduction of PGE2 production but independent of COX-2 expression. We suggest that investigated conjugate may be a promising candidate for therapy of TMZ-resistant glioblastoma multiforme, although applicable in local treatment, since our previous study of G3BC31 did not demonstrate selectivity against glioma cells compared to normal human fibroblasts. However, it has to be pointed that in our in vivo studies conducted with model organism, Caenorhabditis elegans indicated high anti-nematode activity of G3BC31 in comparison with CXB alone that confirms of usefulness of that organism for estimation of anti-cancer drug toxicity.
Sigma-Aldrich
Phenazine methosulfate
Supelco
Temozolomide, Pharmaceutical Secondary Standard; Certified Reference Material