Merck
  • Altered expression of fibroblast activation protein-α (FAP) in colorectal adenoma-carcinoma sequence and in lymph node and liver metastases.
Altered expression of fibroblast activation protein-α (FAP) in colorectal adenoma-carcinoma sequence and in lymph node and liver metastases.
Aging (2020-05-20)
Jon Danel Solano-Iturri, Maider Beitia, Peio Errarte, Julio Calvete-Candenas, María C Etxezarraga, Alberto Loizate, Enrique Echevarria, Iker Badiola, Gorka Larrinaga
ABSTRACT
Colorectal cancer (CRC) is a major health problem in elderly people because of its high incidence and high mortality rate. Despite early screening programs, more than half of CRC patients are diagnosed at advanced stages. Fibroblast activation protein-α (FAP) expression in cancer-associated fibroblasts (CAFs) has been associated with a higher risk of metastases and poor survival. Here, we have analyzed the immunohistochemical expression of FAP in 41 adenoma-carcinoma sequences. In addition, FAP expression was analyzed individually and in combination with β-catenin (BCAT), CD44 and Cyclin-D1 expression in primary tumors and in their corresponding lymph node and liver metastases (n=294). Finally, soluble FAP (sFAP) levels in plasma from CRC patients (n=127) were also analyzed by ELISA. FAP was expressed only in CRC tissue and its expression level was found to be higher in tumors exhibiting deeper local invasion and poorer cancer cell differentiation. FAP and concomitant nuclear BCAT expression in cancer cells at the infiltrating front of primary tumors and in lymph node metastases was independently associated with 5- and 10-year cancer specific and disease-free survival. Moreover, lower sFAP levels correlated with poorer survival. These findings support the potential importance of FAP as a biomarker of CRC development and progression.