Merck
  • Targeting the cytoskeleton to direct pancreatic differentiation of human pluripotent stem cells.
Targeting the cytoskeleton to direct pancreatic differentiation of human pluripotent stem cells.
Nature biotechnology (2020-02-26)
Nathaniel J Hogrebe, Punn Augsornworawat, Kristina G Maxwell, Leonardo Velazco-Cruz, Jeffrey R Millman
ABSTRACT
Generation of pancreatic β cells from human pluripotent stem cells (hPSCs) holds promise as a cell replacement therapy for diabetes. In this study, we establish a link between the state of the actin cytoskeleton and the expression of pancreatic transcription factors that drive pancreatic lineage specification. Bulk and single-cell RNA sequencing demonstrated that different degrees of actin polymerization biased cells toward various endodermal lineages and that conditions favoring a polymerized cytoskeleton strongly inhibited neurogenin 3-induced endocrine differentiation. Using latrunculin A to depolymerize the cytoskeleton during endocrine induction, we developed a two-dimensional differentiation protocol for generating human pluripotent stem-cell-derived β (SC-β) cells with improved in vitro and in vivo function. SC-β cells differentiated from four hPSC lines exhibited first- and second-phase dynamic glucose-stimulated insulin secretion. Transplantation of islet-sized aggregates of these cells rapidly reversed severe preexisting diabetes in mice at a rate close to that of human islets and maintained normoglycemia for at least 9 months.
Sigma-Aldrich
Latrunculin A, from sea sponge, ≥85% (HPLC), waxy solid
Sigma-Aldrich
Cytochalasin D, Ready Made Solution, from Zygosporium mansonii, 5 mg/mL in DMSO
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
α/β Integrin-mediated Cell Adhesion Array Combo Kit, colorimetic, The Alpha/Beta Integrin-Mediated Cell Adhesion Array Kit can be used for assessing the presence or absence of specific integrins on the cell surface.