Merck
  • Degradable Controlled Release Fertilizer Composite Prepared via Extrusion: Fabrication, Characterization, and Release Mechanisms.
Degradable Controlled Release Fertilizer Composite Prepared via Extrusion: Fabrication, Characterization, and Release Mechanisms.
Polymers (2020-02-07)
Siwen Bi, Vincenzo Barinelli, Margaret J Sobkowicz
ABSTRACT
In this work, biodegradable polymers were melt compounded with urea phosphate to fabricate "smart fertilizers" for sustainable agriculture. Urea phosphate (UP) is typically applied as a water-soluble fertilizer to treat phosphorus deficiency in high pH soils. Due to the low diffusion rate of phosphate through slow-release fertilizer coatings, phosphate supply has been considered the "bottleneck" for nitrogen-phosphorous-potassium (NPK) nutrients supply. We study the influence of polymer matrix structure on release kinetics in deionized water using novel polyesters including poly (hexamethylene succinate) (PHS), poly (30% butylene succinate-co-70% hexamethylene succinate) (PBHS 30/70), and PBHS 70/30. Melt processed composites of UP and polyester were analyzed to determine UP loading efficiency and dispersion and distribution of the salt in the polymer matrix. A combined empirical model involving diffusion and erosion mechanisms was found have a good agreement with the experimental release curve. This work provides a solution for environmentally friendly controlled release phosphate fertilizer with good release performance using bio-based and biodegradable polymers.