Merck
  • Intraspecific variation in sensitivity to food availability and temperature-induced phenotypic plasticity in the rotifer Keratella cochlearis.
Intraspecific variation in sensitivity to food availability and temperature-induced phenotypic plasticity in the rotifer Keratella cochlearis.
The Journal of experimental biology (2020-02-29)
Eloísa Ramos-Rodríguez, Emilio Moreno, José María Conde-Porcuna
ABSTRACT
Organisms with wide environmentally induced morphological plasticity and cosmopolitan distribution, e.g. the common freshwater rotifer Keratella cochlearis, are ideal models to study the evolution of plastic polymorphisms and the capacity of zooplankton to adapt to local selection conditions. We investigated population-level differences (population-by-environment interaction) in sensitivity to food availability and temperature-induced phenotypic plasticity between two clones of K. cochlearis isolated from neighboring populations in Ruidera Natural Park (Spain) with different trophic statuses: Tinaja lake (mesotrophic) and Cueva Morenilla lake (eutrophic). Using common-garden experiments, each clone proved to have a different sensitivity to food availability, with substantial phenotypic differences between them. When rotifers grew at moderate temperature (15.6°C), low food levels were more efficiently used by the Tinaja versus Cueva Morenilla clone, whereas high food levels were more efficiently used by the Cueva Morenilla versus Tinaja clone. The posterior spine was much longer and the lorica wider in the Tinaja versus Cueva Morenilla clone, with no difference in lorica length. Phylogenetic analysis based on cytochrome c oxidase subunit I (COI) gene sequences showed that the two populations have the same haplotype. This is the first study to show possible local adaptation by a rotifer species to habitats that consistently differ in food availability. We also detected an intriguing deviation from the expected negative relationship between posterior spine length and temperature. Our experimental results indicate that intermediate temperatures may activate the gene responsible for spine elongation in K. cochlearis This suggests that rotifers in nature could use water temperature as proxy signal of a change in predation risk before defense is needed.