Merck
  • Highly Porous Hybrid Metal-Organic Nanoparticles Loaded with Gemcitabine Monophosphate: a Multimodal Approach to Improve Chemo- and Radiotherapy.
Highly Porous Hybrid Metal-Organic Nanoparticles Loaded with Gemcitabine Monophosphate: a Multimodal Approach to Improve Chemo- and Radiotherapy.
ChemMedChem (2019-11-26)
Xue Li, Erika Porcel, Mario Menendez-Miranda, Jingwen Qiu, Xiaomin Yang, Christian Serre, Alexandra Pastor, Didier Desmaële, Sandrine Lacombe, Ruxandra Gref
ABSTRACT
Nanomedicine recently emerged as a novel strategy to improve the performance of radiotherapy. Herein we report the first application of radioenhancers made of nanoscale metal-organic frameworks (nanoMOFs), loaded with gemcitabine monophosphate (Gem-MP), a radiosensitizing anticancer drug. Iron trimesate nanoMOFs possess a regular porous structure with oxocentered Fe trimers separated by around 5 Å (trimesate linkers). This porosity is favorable to diffuse the electrons emitted from nanoMOFs due to activation by γ radiation, leading to water radiolysis and generation of hydroxyl radicals which create nanoscale damages in cancer cells. Moreover, nanoMOFs act as "Trojan horses", carrying their Gem-MP cargo inside cancer cells to interfere with DNA repair. By displaying different mechanisms of action, both nanoMOFs and incorporated Gem-MP contribute to improve radiation efficacy. The radiation enhancement factor of Gem-MP loaded nanoMOFs reaches 1.8, one of the highest values ever reported. These results pave the way toward the design of engineered nanoparticles in which each component plays a role in cancer treatment by radiotherapy.
Sigma-Aldrich
1-Adamantaneacetic acid, 98%
Pricing and availability is not currently available.
Supelco
Acetic acid – triethylamine solution 1:1, suitable for HPLC, 2M:2M aqueous solution, LiChropur
Pricing and availability is not currently available.