- Improvement of Fatty Acid Profile in Durum Wheat Breads Supplemented with Portulaca oleracea L. Quality Traits of Purslane-Fortified Bread.
Improvement of Fatty Acid Profile in Durum Wheat Breads Supplemented with Portulaca oleracea L. Quality Traits of Purslane-Fortified Bread.
The addition of functional ingredients to breads could have effects on preventing cardiovascular diseases, cancers and inflammation. The incorporation of 0-5-10-15% of three populations of dried purslane flour on the rheological, sensorial and nutritional quality of fortified durum wheat breads were evaluated. The increase in dried purslane (up to 15%) caused an increase in the resistance to the mixture and a consequent reduction in its extensibility. The "panel test" gave a largely positive evaluation in 10% of enrichment. The fatty acids in breads resulted higher with the 5% substitution. Contrary to what has been imagined, the increase in percentage of substitution to 10 and 15% did not lead to an increase in linoleic (omega-3) and α-linolenic (omega-6) acid and probably the cause is in the cooking. The total phenols content and the antioxidant potential, evaluated by ferric reducing antioxidant potential (FRAP) and 2,2'-azinobis (3-ethylbenzothiazoline-6-sulfonic acid) (ABTS) assays of the enriched breads increased with the percentage of the dry purslane substitution. The enrichment of the durum wheat flour with 5% purslane resulted in a good compromise to obtain good rheological characteristics of loaves and breads with decreased omega-6/omega-3 ratio and good antioxidant properties.