Merck
  • The Ubiquitin Ligase TRIP12 Limits PARP1 Trapping and Constrains PARP Inhibitor Efficiency.
The Ubiquitin Ligase TRIP12 Limits PARP1 Trapping and Constrains PARP Inhibitor Efficiency.
Cell reports (2020-08-07)
Marco Gatti, Ralph Imhof, Qingyao Huang, Michael Baudis, Matthias Altmeyer
ABSTRACT
PARP inhibitors (PARPi) cause synthetic lethality in BRCA-deficient tumors. Whether specific vulnerabilities to PARPi exist beyond BRCA mutations and related defects in homology-directed repair (HDR) is not well understood. Here, we identify the ubiquitin E3 ligase TRIP12 as negative regulator of PARPi sensitivity. We show that TRIP12 controls steady-state PARP1 levels and limits PARPi-induced cytotoxic PARP1 trapping. Upon loss of TRIP12, elevated PARPi-induced PARP1 trapping causes increased DNA replication stress, DNA damage, cell cycle arrest, and cell death. Mechanistically, we demonstrate that TRIP12 binds PARP1 via a central PAR-binding WWE domain and, using its carboxy-terminal HECT domain, catalyzes polyubiquitylation of PARP1, triggering proteasomal degradation and preventing supra-physiological PARP1 accumulation. Further, in cohorts of breast and ovarian cancer patients, PARP1 abundance is negatively correlated with TRIP12 expression. We thus propose TRIP12 as regulator of PARP1 stability and PARPi-induced PARP trapping, with potential implications for PARPi sensitivity and resistance.
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
2′-Deoxyadenosine 5′-triphosphate sodium salt solution, 100 mM, pH 7
Sigma-Aldrich
3X FLAG® Peptide, lyophilized powder
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
2′-Deoxyguanosine 5′-triphosphate trisodium salt solution, 100 mM (pH 7)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-53BP1 Antibody, clone BP13, clone BP13, Chemicon®, from mouse
Roche
Rapid DNA Ligation Kit
Sigma-Aldrich
FK866 hydrochloride hydrate, ≥98% (HPLC)
Sigma-Aldrich
Methyl methanesulfonate, 99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
2′-Deoxycytidine 5′-triphosphate disodium salt, 100 mM (pH 7)
Sigma-Aldrich
MG-132, Ready Made Solution, ≥90% (HPLC)