Merck
  • siRNA nanoparticles targeting CaMKIIγ in lesional macrophages improve atherosclerotic plaque stability in mice.
siRNA nanoparticles targeting CaMKIIγ in lesional macrophages improve atherosclerotic plaque stability in mice.
Science translational medicine (2020-07-29)
Wei Tao, Arif Yurdagul, Na Kong, Wenliang Li, Xiaobo Wang, Amanda C Doran, Chan Feng, Junqing Wang, Mohammad Ariful Islam, Omid C Farokhzad, Ira Tabas, Jinjun Shi
ABSTRACT
Atherosclerotic lesional macrophages express molecules that promote plaque progression, but lack of mechanisms to therapeutically target these molecules represents a major gap in translational cardiovascular research. Here, we tested the efficacy of a small interfering RNA (siRNA) nanoparticle (NP) platform targeting a plaque-destabilizing macrophage molecule-Ca2+/calmodulin-dependent protein kinase γ (CaMKIIγ). CaMKIIγ becomes activated in advanced human and mouse plaque macrophages and drives plaque necrosis by suppressing the expression of the efferocytosis receptor MerTK. When macrophage-targeted siCamk2g NPs were administered to Western diet-fed Ldlr-/- mice, the atherosclerotic lesions showed decreased CaMKIIγ and increased MerTK expression in macrophages, improved phagocytosis of apoptotic cells (efferocytosis), decreased necrotic core area, and increased fibrous cap thickness-all signs of increased plaque stability-compared with mice treated with control siRNA NPs. These findings demonstrate that atherosclerosis-promoting genes in plaque macrophages can be targeted with siRNA NPs in a preclinical model of advanced atherosclerosis.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, for general use, Type I, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type XI, 2-5 FALGPA units/mg solid, ≥800 CDU/mg solid
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid