Merck
  • Temporal pressure enhanced topical drug delivery through micropore formation.
Temporal pressure enhanced topical drug delivery through micropore formation.
Science advances (2020-06-12)
Daniel Chin Shiuan Lio, Rui Ning Chia, Milton Sheng Yi Kwek, Christian Wiraja, Leigh Edward Madden, Hao Chang, S Mohideen Abdul Khadir, Xiaomeng Wang, David L Becker, Chenjie Xu
ABSTRACT
Transdermal drug delivery uses chemical, physical, or biochemical enhancers to cross the skin barrier. However, existing platforms require high doses of chemical enhancers or sophisticated equipment, use fragile biomolecules, or are limited to a certain type of drug. Here, we report an innovative methodology based on temporal pressure to enhance the penetration of all kinds of drugs, from small molecules to proteins and nanoparticles (up to 500 nm). The creation of micropores (~3 μm2) on the epidermal layer through a temporal pressure treatment results in the elevated expression of gap junctions, and reduced expression of occludin tight junctions. A 1 min treatment of 0.28-MPa allows nanoparticles (up to 500 nm) and macromolecules (up to 20 kDa) to reach a depth of 430-μm into the dermal layer. Using, as an example, the delivery of insulin through topical application after the pressure treatment yields up to 80% drop in blood glucose in diabetic mice.
Sigma-Aldrich
Organo/Limonene Mount