Merck
  • Unrestrained ESCRT-III drives micronuclear catastrophe and chromosome fragmentation.
Unrestrained ESCRT-III drives micronuclear catastrophe and chromosome fragmentation.
Nature cell biology (2020-07-01)
Marina Vietri, Sebastian W Schultz, Aurélie Bellanger, Carl M Jones, Louise I Petersen, Camilla Raiborg, Ellen Skarpen, Christeen Ramane J Pedurupillay, Ingrid Kjos, Eline Kip, Romy Timmer, Ashish Jain, Philippe Collas, Roland L Knorr, Sushma N Grellscheid, Halim Kusumaatmaja, Andreas Brech, Francesca Micci, Harald Stenmark, Coen Campsteijn
ABSTRACT
The ESCRT-III membrane fission machinery maintains the integrity of the nuclear envelope. Although primary nuclei resealing takes minutes, micronuclear envelope ruptures seem to be irreversible. Instead, micronuclear ruptures result in catastrophic membrane collapse and are associated with chromosome fragmentation and chromothripsis, complex chromosome rearrangements thought to be a major driving force in cancer development. Here we use a combination of live microscopy and electron tomography, as well as computer simulations, to uncover the mechanism underlying micronuclear collapse. We show that, due to their small size, micronuclei inherently lack the capacity of primary nuclei to restrict the accumulation of CHMP7-LEMD2, a compartmentalization sensor that detects loss of nuclear integrity. This causes unrestrained ESCRT-III accumulation, which drives extensive membrane deformation, DNA damage and chromosome fragmentation. Thus, the nuclear-integrity surveillance machinery is a double-edged sword, as its sensitivity ensures rapid repair at primary nuclei while causing unrestrained activity at ruptured micronuclei, with catastrophic consequences for genome stability.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
S-Nitroso-N-acetyl-DL-penicillamine, ≥97%, powder
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid