Merck
  • Cytoplasm lipids can be modulated through hormone-sensitive lipase and are related to mitochondrial function in porcine IVM oocytes.
Cytoplasm lipids can be modulated through hormone-sensitive lipase and are related to mitochondrial function in porcine IVM oocytes.
Reproduction, fertility, and development (2020-03-17)
Qingrui Zhuan, Haojia Ma, Jing Chen, Yuxi Luo, Yan Luo, Lei Gao, Yunpeng Hou, Shien Zhu, Xiangwei Fu
ABSTRACT
Intracellular lipids provide energy for oocyte maturation and development. Triglycerides are the main components of cytoplasm lipid droplets, and hydrolysis of triglycerides requires several lipase-mediated steps. The aim of this study was to determine the effects of the β-adrenoceptor agonist isoproterenol (ISO) and the hormone-sensitive lipase (HSL) inhibitor CAY10499 on the IVM of porcine oocytes. ISO (5mg L-1) and CAY10499 (20mg L-1) had positive and negative effects respectively on invitro oocyte maturation and subsequent embryo development. The rates of polar body extrusion, cleavage and blastocyst formation were significantly higher in the ISO-treated group than the control and CAY10499-treated groups. ISO treatment also upregulated intracellular cAMP levels in comparison with the control group, while CAY10499 significantly increased the triglyceride content of matured oocytes when compared with other groups, consistent with the observed decrease in LIPE (HSL) mRNA levels. Furthermore, the inhibitory effects of CAY10499 included decreases in mitochondrial membrane potential and mitochondrial temperature. These results indicate that ISO has a positive effect on the IVM of porcine oocytes, and that intracellular lipid metabolism can be modulated by CAY10499 through inhibition of HSL and is closely related to mitochondrial function.
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Pricing and availability is not currently available.
Sigma-Aldrich
Triglycerol, technical
Pricing and availability is not currently available.
Sigma-Aldrich
Triglyceride Reagent, used for quantitative enzymatic determination of triglyerides
Pricing and availability is not currently available.