Merck
  • Systematic characterization of glutathione S-transferases in common marmosets.
Systematic characterization of glutathione S-transferases in common marmosets.
Biochemical pharmacology (2020-02-07)
Yasuhiro Uno, Shotaro Uehara, Saki Tanaka, Norie Murayama, Hiroshi Yamazaki
ABSTRACT
The common marmoset is an important primate species used in drug metabolism studies. However, glutathione S-transferases (GSTs), essential drug-metabolizing enzymes involved in the conjugation of various endogenous and exogenous substrates, have not been identified or characterized in this species. In this study, 20 GSTs [including 3 microsomal GSTs (MGSTs)] were identified and characterized in marmosets. Marmoset GSTs had amino acid sequences highly identical (86-99%) to human GSTs, except for GSTA4L, which had lower identities (59-62%) with human GSTAs. Phylogenetic analysis revealed that marmoset GSTs were closely clustered with their human counterparts. Marmoset GSTs had gene and genomic structures generally similar to their human counterparts, with some differences in GSTA, GSTM, and GSTT clusters. Marmoset GST mRNAs exhibited distinct tissue expression patterns: GSTA1, GSTA3, GSTA4L, GSTK1, GSTT1, GSTZ1, and MGST1 mRNAs were expressed most abundantly in liver. Other GST mRNAs were expressed most abundantly in small intestine, lung, brain, or kidney. Expression of GSTT4 and GSTT4L mRNAs was detected only in testis. Among all 20 marmoset GST mRNAs, the most abundant mRNAs were GSTA1 mRNA in liver, small intestine, and kidney; GSTM3 mRNA in testis; and MSGT3 mRNA in brain and lung. All 20 GSTs mediated the conjugation of GST substrates 1-chloro-2,4-dinitrobenzene; 1,2-epoxy-3-(p-nitrophenoxy)propane; styrene 7,8-oxide; and/or 1-iodohexane, but with different activity levels. Kinetic analyses showed that marmoset GSTM2/GSTM5 and GSTM5/GSTT1 effectively conjugated styrene 7,8-oxide and 1-iodohexane, respectively, with the highest affinity. These results suggest that the 20 newly identified marmoset GSTs were functional drug-metabolizing enzymes able to conjugate typical GST substrates.