Merck
  • ZNF143-Mediated H3K9 Trimethylation Upregulates CDC6 by Activating MDIG in Hepatocellular Carcinoma.
ZNF143-Mediated H3K9 Trimethylation Upregulates CDC6 by Activating MDIG in Hepatocellular Carcinoma.
Cancer research (2020-04-22)
Lili Zhang, Qi Huo, Chao Ge, Fangyu Zhao, Qingqing Zhou, Xiaoxia Chen, Hua Tian, Taoyang Chen, Haiyang Xie, Ying Cui, Ming Yao, Hong Li, Jinjun Li
ABSTRACT
Zinc finger protein 143 (ZNF143) belongs to the zinc finger protein family and possesses transcription factor activity by binding sequence-specific DNA. The exact biological role of ZNF143 in hepatocellular carcinoma (HCC) has not been investigated. Here we report that ZNF143 is overexpressed in HCC tissues and its overexpression correlates with poor prognosis. Gain- and loss-of-function experiments showed that ZNF143 promoted HCC cell proliferation, colony formation, and tumor growth in vitro and in vivo. ZNF143 accelerated HCC cell-cycle progression by activating cell division cycle 6 (CDC6). Mechanistically, ZNF143 promoted expression of CDC6 by directly activating transcription of histone demethylase mineral dust-induced gene (MDIG), which in turn reduced H3K9me3 enrichment in the CDC6 promoter region. Consistently, ZNF143 expression correlated significantly with MDIG and CDC6 expression in HCC. Collectively, we propose a model for a ZNF143-MDIG-CDC6 oncoprotein axis that provides novel insight into ZNF143, which may serve as a therapeutic target in HCC. SIGNIFICANCE: These findings describe the mechanism by which ZNF143 promotes HCC proliferation and provide important clues for exploring new targets and strategies for clinical treatment of human liver cancer.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Telbivudine, ≥98% (HPLC)