Merck
  • RAI1 Regulates Activity-Dependent Nascent Transcription and Synaptic Scaling.
RAI1 Regulates Activity-Dependent Nascent Transcription and Synaptic Scaling.
Cell reports (2020-08-14)
Patricia M Garay, Alex Chen, Takao Tsukahara, Jean Carlos Rodríguez Díaz, Rafi Kohen, J Christian Althaus, Margarete A Wallner, Roman J Giger, Kevin S Jones, Michael A Sutton, Shigeki Iwase
ABSTRACT
Long-lasting forms of synaptic plasticity such as synaptic scaling are critically dependent on transcription. Activity-dependent transcriptional dynamics in neurons, however, remain incompletely characterized because most previous efforts relied on measurement of steady-state mRNAs. Here, we use nascent RNA sequencing to profile transcriptional dynamics of primary neuron cultures undergoing network activity shifts. We find pervasive transcriptional changes, in which ∼45% of expressed genes respond to network activity shifts. We further link retinoic acid-induced 1 (RAI1), the Smith-Magenis syndrome gene, to the transcriptional program driven by reduced network activity. Remarkable agreement among nascent transcriptomes, dynamic chromatin occupancy of RAI1, and electrophysiological properties of Rai1-deficient neurons demonstrates the essential roles of RAI1 in suppressing synaptic upscaling in the naive network, while promoting upscaling triggered by activity silencing. These results highlight the utility of bona fide transcription profiling to discover mechanisms of activity-dependent chromatin remodeling that underlie normal and pathological synaptic plasticity.
Sigma-Aldrich
MISSION® TRC2 pLKO.5-puro Non-Mammalian shRNA Control Plasmid DNA, Targets no known mammalian genes
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Anti-GluR1 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, Peroxidase Conjugated, 1 mg/mL (after reconstitution), Chemicon®
Sigma-Aldrich
Anti-MAP2 Antibody, Chemicon®, from chicken
Sigma-Aldrich
5-Bromouridine, 98%
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Rabbit Anti-Mouse IgG Antibody, F(ab′)2, HRP conjugate, Chemicon®, from rabbit
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Post Synaptic Density Protein 95 Antibody, clone 6G6-1C9, clone 6G6-1C9, Chemicon®, from mouse
Sigma-Aldrich
TRI Reagent®, BD, For processing whole blood, plasma, or serum.