Merck
Palladium-catalyzed oxygenation of unactivated sp3 C-H bonds.
Journal of the American Chemical Society (2004-08-05)
Lopa V Desai, Kami L Hull, Melanie S Sanford
ABSTRACT
This communication describes a new palladium-catalyzed method for the oxygenation of unactivated sp3 C-H bonds. A wide variety of alkane substrates containing readily available oxime and/or pyridine directing groups are oxidized with extremely high levels of chemo-, regio-, and in some cases diastereoselectivity. The substrate scope of these reactions is discussed, and the high selectivities are rationalized on the basis of the requirements of putative palladium alkyl intermediates.
Sigma-Aldrich
(Diacetoxyiodo)benzene, 98%
Pricing and availability is not currently available.