- PLAGA-PEG-PLAGA Terpolymer-Based Carriers of Herbicides for Potential Application in Environment-Friendly, Controlled Release Systems of Agrochemicals.
PLAGA-PEG-PLAGA Terpolymer-Based Carriers of Herbicides for Potential Application in Environment-Friendly, Controlled Release Systems of Agrochemicals.
The present study aimed to develop and prepare new polymer/herbicide formulations for their potential application in environment-friendly, controlled release systems of agrochemicals. Selected biodegradable polymers, including L-Lactide/Glycolide/PEG/Terpolymer (PLAGA-PEG-PLAGA) as well as oligosaccharide-based polymers and their blend with terpolymer, were used to prepare microspheres loaded with two soil-applied herbicides. The degradation process of the obtained polymeric microspheres was evaluated based on (1) their weight loss and surface erosion and (2) the release rate of loaded metazachlor and pendimethalin. The herbicidal effectiveness of the herbicides released to the soil from microspheres was evaluated using the European Weed Research Council (EWRC) rating scale. Moreover, the ecotoxicological effect of herbicide-loaded microspheres buried in soil on the marine bacterial species A. fischeri was assessed. It was found that the gradual degradation rate of microparticles led to the prolonged release of both herbicides that lasted for a few months, i.e., for the entire crop season, which is crucial in terms of agrochemical and environmental protection. Maltodextrin- and dextrin-based microspheres showed higher susceptibility to degradation than terpolymer-based microspheres. The microencapsulation of herbicides protected them from decomposition and excessive leaching into soil and maintained their activity for a longer period than that for non-immobilized herbicides. The ecotoxicological assessment on A. fischeri demonstrated that the proposed microsphere-encapsulated herbicides were less toxic than non-immobilized herbicides.