Merck
  • Multiple low dose therapy as an effective strategy to treat EGFR inhibitor-resistant NSCLC tumours.
Multiple low dose therapy as an effective strategy to treat EGFR inhibitor-resistant NSCLC tumours.
Nature communications (2020-06-24)
João M Fernandes Neto, Ernest Nadal, Evert Bosdriesz, Salo N Ooft, Lourdes Farre, Chelsea McLean, Sjoerd Klarenbeek, Anouk Jurgens, Hannes Hagen, Liqin Wang, Enriqueta Felip, Alex Martinez-Marti, August Vidal, Emile Voest, Lodewyk F A Wessels, Olaf van Tellingen, Alberto Villanueva, René Bernards
ABSTRACT
Resistance to targeted cancer drugs is thought to result from selective pressure exerted by a high drug dose. Partial inhibition of multiple components in the same oncogenic signalling pathway may add up to complete pathway inhibition, while decreasing the selective pressure on each component to acquire a resistance mutation. We report here testing of this Multiple Low Dose (MLD) therapy model in EGFR mutant NSCLC. We show that as little as 20% of the individual effective drug doses is sufficient to completely block MAPK signalling and proliferation when used in 3D (RAF + MEK + ERK) or 4D (EGFR + RAF + MEK + ERK) inhibitor combinations. Importantly, EGFR mutant NSCLC cells treated with MLD therapy do not develop resistance. Using several animal models, we find durable responses to MLD therapy without associated toxicity. Our data support the notion that MLD therapy could deliver clinical benefit, even for those having acquired resistance to third generation EGFR inhibitor therapy.
Sigma-Aldrich
LY3009120, ≥98% (HPLC)
SAFC
Formaldehyde solution, 37%, EMPROVE® EVOLVE
Sigma-Aldrich
Dispase® II, protease
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid