Merck
  • Reduced circulating BMP10 and BMP9 and elevated endoglin are associated with disease severity, decompensation and pulmonary vascular syndromes in patients with cirrhosis.
Reduced circulating BMP10 and BMP9 and elevated endoglin are associated with disease severity, decompensation and pulmonary vascular syndromes in patients with cirrhosis.
EBioMedicine (2020-05-27)
Nicola E Owen, Graeme J Alexander, Sambit Sen, Katherine Bunclark, Gary Polwarth, Joanna Pepke-Zaba, Anthony P Davenport, Nicholas W Morrell, Paul D Upton
ABSTRACT
BMP9, originating from the liver, and BMP10 are circulating BMPs that preserve vascular endothelial integrity. We assessed BMP9, BMP10 and soluble endoglin (sEng) levels and their relationships to liver disease severity and associated pulmonary vascular syndromes in a cohort of well-characterised liver disease patients. Plasma samples from patients with liver disease (n = 83) and non-disease controls (n = 21) were assayed for BMP9, BMP10 and sEng. Levels were also assessed in a separate cohort of controls (n = 27) and PoPH patients (n = 8). Expression of mRNA and immunohistochemical staining was undertaken in liver biopsy specimens. Plasma BMP activity was assessed using an endothelial cell bioassay. Plasma BMP9 and BMP10 levels were normal in patients with compensated cirrhosis or fibrosis without cirrhosis, but markedly reduced in patients with decompensated cirrhosis, including those with hepatopulmonary syndrome (HPS) or portopulmonary hypertension (PoPH). Liver biopsy specimens revealed reduced mRNA expression and immunostaining for these ligands. Patient plasma samples with reduced BMP9 and BMP10 levels exhibited low BMP activity that was restored with exogenous BMP9. Endoglin mRNA expression was increased in cirrhotic livers and elevated circulating sEng levels in PoPH and HPS patients suggested increased endothelial sEng shedding in these syndromes. Plasma BMP9 and BMP10 levels are reduced in decompensated cirrhosis, leading to reduced circulating BMP activity on the vascular endothelium. The pulmonary complications of cirrhosis, PoPH and HPS, are associated with markedly reduced BMP9 and BMP10 and increased sEng levels, suggesting that supplementation with exogenous ligands might be a therapeutic approach for PoPH and HPS.
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)