Merck
  • Antagonistic Activities of Vegfr3/Flt4 and Notch1b Fine-tune Mechanosensitive Signaling during Zebrafish Cardiac Valvulogenesis.
Antagonistic Activities of Vegfr3/Flt4 and Notch1b Fine-tune Mechanosensitive Signaling during Zebrafish Cardiac Valvulogenesis.
Cell reports (2020-07-16)
Federica Fontana, Timm Haack, Maria Reichenbach, Petra Knaus, Michel Puceat, Salim Abdelilah-Seyfried
ABSTRACT
The formation of cardiac valves depends on mechanical forces exerted by blood flow. Endocardial cells lining the interior of the heart are sensitive to these stimuli and respond by rearranging into luminal cells subjected to shear stress and abluminal cells not exposed to it. The mechanisms by which endocardial cells sense these dynamic biomechanical stimuli and how they evoke different cellular responses are largely unknown. Here, we show that blood flow activates two parallel mechanosensitive pathways, one mediated by Notch and the other by Klf2a. Both pathways negatively regulate the angiogenesis receptor Vegfr3/Flt4, which becomes restricted to abluminal endocardial cells. Its loss disrupts valve morphogenesis and results in the occurrence of Notch signaling within abluminal endocardial cells. Our work explains how antagonistic activities by Vegfr3/Flt4 on the abluminal side and by Notch on the luminal side shape cardiac valve leaflets by triggering unique differences in the fates of endocardial cells.
Sigma-Aldrich
5-Propyl-2-thiouracil, ≥98%
Roche
KAPA SYBR® FAST One-Step, suitable for RT-PCR, 2 ×, Universal
Sigma-Aldrich
Myokinase from rabbit muscle, ammonium sulfate suspension, 1,500-3,000 units/mg protein (biuret)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)