Merck
  • MicroRNA-3690 promotes cell proliferation and cell cycle progression by altering DKK3 expression in human thyroid cancer.
MicroRNA-3690 promotes cell proliferation and cell cycle progression by altering DKK3 expression in human thyroid cancer.
Oncology letters (2020-09-25)
Fei Shen, Xiao-Xiong Gan, Xing-Yan Deng, Jian-Hua Feng, Wen-Song Cai, Liang Shen, Huan-Qing Xiao, Bo Xu
ABSTRACT
An increasing amount of evidence has demonstrated the importance of microRNAs (miRNAs/miRs) in the tumorigenesis of malignant types of cancer, and data retrieved from The Cancer Genome Atlas database revealed that miR-3690 was upregulated in thyroid cancer (TC). The present study focused on the biological function and mechanism of miR-3690 in TC, demonstrating that miR-3690 expression was significantly elevated in TC cells and clinical tissues. Functional studies indicated that miR-3690 acted as an oncogene in TC by promoting cell proliferation, colony formation and cell cycle progression in association with the increased expression of cyclin E and c-myc. Mechanistically, prediction software indicated that Dickkopf-related protein 3 (DKK3) was a target of miR-3690, which was confirmed by the results of luciferase reporter assays and western blotting. DKK3 silencing abrogated the functions of miR-3690-in on TC cell proliferation. Collectively, the findings of the present study demonstrated that miR-3690 promoted TC cell proliferation and indicated miR-3690 as a potential biomarker and therapeutic target for TC.