Merck
  • Leukotriene B4 Receptor Type 2 Accelerates the Healing of Intestinal Lesions by Promoting Epithelial Cell Proliferation.
Leukotriene B4 Receptor Type 2 Accelerates the Healing of Intestinal Lesions by Promoting Epithelial Cell Proliferation.
The Journal of pharmacology and experimental therapeutics (2020-01-17)
Yui Matsumoto, Yukiko Matsuya, Kano Nagai, Kikuko Amagase, Kazuko Saeki, Kenjiro Matsumoto, Takehiko Yokomizo, Shinichi Kato
ABSTRACT
Leukotriene B4 receptor type 2 (BLT2) is a low-affinity leukotriene B4 receptor that is highly expressed in intestinal epithelial cells. Previous studies demonstrated the protective role of BLT2 in experimentally induced colitis. However, its role in intestinal lesion repair is not fully understood. We investigated the role of BLT2 in the healing of indomethacin-induced intestinal lesions in mice. There was no significant different between wild-type (WT) and BLT2-deficient (BLT2KO) mice in terms of the development of indomethacin-induced intestinal lesions. However, healing of these lesions was significantly impaired in BLT2KO mice compared with WT mice. In contrast, transgenic mice with intestinal epithelium-specific BLT2 overexpression presented with superior ileal lesion healing relative to WT mice. An immunohistochemical study showed that the number of Ki-67-proliferative cells was markedly increased during the healing of intestinal lesions in WT mice but significantly attenuated in BLT2KO mice. Exposure of cultured mouse intestinal epithelial cells to CAY10583, a BLT2 agonist, promoted wound healing and cell proliferation in a concentration-dependent manner. Nevertheless, these responses were abolished under serum-free conditions. The CAY10583-induced proliferative effect was also negated by Go6983, a protein kinase C (PKC) inhibitor, U-73122, a phospholipase C (PLC) inhibitor, LY255283, a BLT2 antagonist, and pertussis toxin that inhibits G protein-coupled receptor signaling via Gi/o proteins. Thus, BLT2 plays an important role in intestinal wound repair. Moreover, this effect is mediated by the promotion of epithelial cell proliferation via the Gi/o protein-dependent and PLC/PKC signaling pathways. The BLT2 agonists are potential therapeutic agents for the treatment of intestinal lesions. SIGNIFICANCE STATEMENT: The healing of indomethacin-induced Crohn's disease-like intestinal lesions was impaired in mice deficient in low-affinity leukotriene B4 receptor type 2 (BLT2). They presented with reduced epithelial cell proliferation during the healing. In contrast, healing was promoted in mice overexpressing intestinal epithelial BLT2. In cultured intestinal epithelial cells, the BLT2 agonist CAY10583 substantially accelerated wound repair by enhancing cell proliferation rather than migration. Thus, BLT2 plays an important role in the intestinal lesions via acceleration of epithelial cell proliferation.
Sigma-Aldrich
CAY10583, ≥98% (HPLC)