Merck
  • Rapid and Sensitive Detection of Hepatitis C Virus in Clinical Blood Samples Using Reverse Transcriptase Polymerase Spiral Reaction.
Rapid and Sensitive Detection of Hepatitis C Virus in Clinical Blood Samples Using Reverse Transcriptase Polymerase Spiral Reaction.
Journal of microbiology and biotechnology (2020-01-03)
Wenying Sun, Ying Du, Xingku Li, Bo Du
ABSTRACT
This study established a new polymerase spiral reaction (PSR) that combines with reverse transcription reactions for HCV detection targeting 5'UTR gene. To avoid cross-contamination of aerosols, an isothermal amplification tube (IAT), as a separate containment control, was used to judge the result. After optimizing the RT-PSR reaction system, its effectiveness and specificity were tested against 15 different virus strains which included 8 that were HCV positive and 7 as non-HCV controls. The results showed that the RT-PSR assay effectively detected all 8 HCV strains, and no false positives were found among the 7 non-HCV strains. The detection limit of our RT-PSR assay is comparable to the real-time RT-PCR, but is more sensitive than the RT-LAMP. The established RT-PSR assay was further evaluated for detection of HCV in clinical blood samples, and the resulting 80.25% detection rate demonstrated better or similar effectiveness compared to the RT-LAMP (79.63%) and real-time RT-PCR (80.25%). Overall, the results showed that the RT-PSR assay offers high specificity and sensitivity for HCV detection with great potential for screening HCV in clinical blood samples.