Merck
  • Rap1-GTPases control mTORC1 activity by coordinating lysosome organization with amino acid availability.
Rap1-GTPases control mTORC1 activity by coordinating lysosome organization with amino acid availability.
Nature communications (2020-03-19)
Anders P Mutvei, Michal J Nagiec, Jens C Hamann, Sang Gyun Kim, C Theresa Vincent, John Blenis
ABSTRACT
The kinase mTOR complex 1 (mTORC1) promotes cellular growth and is frequently dysregulated in cancers. In response to nutrients, mTORC1 is activated on lysosomes by Rag and Rheb guanosine triphosphatases (GTPases) and drives biosynthetic processes. How limitations in nutrients suppress mTORC1 activity remains poorly understood. We find that when amino acids are limited, the Rap1-GTPases confine lysosomes to the perinuclear region and reduce lysosome abundance, which suppresses mTORC1 signaling. Rap1 activation, which is independent of known amino acid signaling factors, limits the lysosomal surface available for mTORC1 activation. Conversely, Rap1 depletion expands the lysosome population, which markedly increases association between mTORC1 and its lysosome-borne activators, leading to mTORC1 hyperactivity. Taken together, we establish Rap1 as a critical coordinator of the lysosomal system, and propose that aberrant changes in lysosomal surface availability can impact mTORC1 signaling output.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MISSION® esiRNA, targeting human LAMP1
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-(−)-Glucose, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Latrunculin A, from sea sponge, ≥85% (HPLC), waxy solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MISSION® esiRNA, targeting human LAMP2
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture