Merck
  • Increased mitochondrial calcium levels associated with neuronal death in a mouse model of Alzheimer's disease.
Increased mitochondrial calcium levels associated with neuronal death in a mouse model of Alzheimer's disease.
Nature communications (2020-05-03)
Maria Calvo-Rodriguez, Steven S Hou, Austin C Snyder, Elizabeth K Kharitonova, Alyssa N Russ, Sudeshna Das, Zhanyun Fan, Alona Muzikansky, Monica Garcia-Alloza, Alberto Serrano-Pozo, Eloise Hudry, Brian J Bacskai
ABSTRACT
Mitochondria contribute to shape intraneuronal Ca2+ signals. Excessive Ca2+ taken up by mitochondria could lead to cell death. Amyloid beta (Aβ) causes cytosolic Ca2+ overload, but the effects of Aβ on mitochondrial Ca2+ levels in Alzheimer's disease (AD) remain unclear. Using a ratiometric Ca2+ indicator targeted to neuronal mitochondria and intravital multiphoton microscopy, we find increased mitochondrial Ca2+ levels associated with plaque deposition and neuronal death in a transgenic mouse model of cerebral β-amyloidosis. Naturally secreted soluble Aβ applied onto the healthy brain increases Ca2+ concentration in mitochondria, which is prevented by blockage of the mitochondrial calcium uniporter. RNA-sequencing from post-mortem AD human brains shows downregulation in the expression of mitochondrial influx Ca2+ transporter genes, but upregulation in the genes related to mitochondrial Ca2+ efflux pathways, suggesting a counteracting effect to avoid Ca2+ overload. We propose lowering neuronal mitochondrial Ca2+ by inhibiting the mitochondrial Ca2+ uniporter as a novel potential therapeutic target against AD.
Sigma-Aldrich
Anti-SLC24A6 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse