Merck
  • Osmolyte transporter expression is reduced in photoaged human skin: Implications for skin hydration in aging.
Osmolyte transporter expression is reduced in photoaged human skin: Implications for skin hydration in aging.
Aging cell (2019-11-27)
April R Foster, Cecile El Chami, Catherine A O'Neill, Rachel E B Watson
ABSTRACT
Aging is characterized by the deterioration of tissue structure and function. In skin, environmental factors, for example, ultraviolet radiation (UVR), can accelerate the effects of aging such as decline in barrier function and subsequent loss of hydration. Water homeostasis is vital for all cellular functions and it is known that organic osmolyte transport is critical to this process. Therefore, we hypothesized that as we age, these tightly controlled physiological mechanisms become disrupted, possibly due to loss of transporter expression. We investigated this in vivo, using human skin samples from photoprotected and photoexposed sites of young and aged volunteers. We show a reduction in keratinocyte cell size with age and a downregulation of osmolyte transporters SMIT and TAUT with both chronic and acute UVR exposure. Single-cell live imaging demonstrated that aged keratinocytes lack efficient cell volume recovery mechanisms possessed by young keratinocytes following physiological stress. However, addition of exogenous taurine significantly rescued cell volume; this was corroborated by a reduction in TAUT mRNA and protein in aged, as compared to young, keratinocytes. Collectively, these novel data demonstrate that human epidermal keratinocytes possess osmolyte-mediated cell volume regulatory mechanisms, which may be compromised in aging. Therefore, this suggests that organic osmolytes-especially taurine-play a critical role in cutaneous age-related xerosis and highlights a fundamental mechanism, vital to our understanding of the pathophysiology of skin aging.
Sigma-Aldrich
Anti-SLC5A3 Antibody, from rabbit, purified by affinity chromatography