Merck
  • Data for engineering lipid metabolism of Chinese hamster ovary (CHO) cells for enhanced recombinant protein production.
Data for engineering lipid metabolism of Chinese hamster ovary (CHO) cells for enhanced recombinant protein production.
Data in brief (2020-02-20)
James D Budge, Tanya J Knight, Jane Povey, Joanne Roobol, Ian R Brown, Gurdeep Singh, Andrew Dean, Sarah Turner, Colin M Jaques, Robert J Young, Andrew J Racher, C Mark Smales
ABSTRACT
The data presented in this article relates to the manuscript entitled 'Engineering of Chinese hamster ovary cell lipid metabolism results in an expanded ER and enhanced recombinant biotherapeutic protein production', published in the Journal Metabolic Engineering [1]. In the article here, we present data examining the overexpression of the lipid metabolism modifying genes SCD1 and SREBF1 in CHO cells by densitometry of western blots and by using mass spectrometry to investigate the impact on specific lipid species. We also present immunofluorescence data at the protein level upon SCD1 and SREBF1 overexpression. The growth profile data during batch culture of control CHO cells and CHO cells engineered to overexpress SCD1 and SREBF1 during batch culture are also reported. Finally, we report data on the yields of model secretory recombinant proteins produced from control, SCD1 or SREBF1 engineered cells using a transient expression systems.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution