Merck
  • Deciphering archaeal glycolipids of an extremely halophilic archaeon of the genus Halobellus by MALDI-TOF/MS.
Deciphering archaeal glycolipids of an extremely halophilic archaeon of the genus Halobellus by MALDI-TOF/MS.
Chemistry and physics of lipids (2014-12-03)
Simona Lobasso, Azahara Pérez-Davó, Rita Vitale, Mercedes Monteoliva- Sánchez, Angela Corcelli
ABSTRACT
Polar membrane lipids of an archaeal microorganism recently isolated from the natural salt lake Fuente de Piedra (Málaga, Spain) have been studied by means of TLC in combination with MALDI-TOF mass spectrometry. The major phospholipids are the ether lipids phosphatidylglycerophosphate methyl ester and phosphatidylglycerosulfate, while phosphatidylglycerol is barely detectable; in addition the bisphosphatidylglycerol (archaeal cardiolipin) has been detected for the first time in a representative of the genus Halobellus. The structures of glycolipids, including a glycosyl-cardiolipin, have been elucidated by post source decay (PSD) mass spectrometry analysis. Besides the monosulfated diglycosyl diphytanylglyceroldiether, two variants of a bis-sulfated diglycosyl diphytanylglyceroldiether have been identified; furthermore the glycosyl-cardiolipin is found to have the same structure of the analogue present in Halorubrum trapanicum and Haloferax volcanii. The role of the abundant sulfated glycolipids in facing high extracellular salinity is discussed.
Sigma-Aldrich
Molybdenum Blue spray reagent