Merck
  • Functional characterization of wild-type and 24 CYP2D6 allelic variants on gefitinib metabolism in vitro.
Functional characterization of wild-type and 24 CYP2D6 allelic variants on gefitinib metabolism in vitro.
Drug design, development and therapy (2017-05-04)
Ping Fang, Xiang Zheng, Jiayang He, Honglei Ge, Pengfei Tang, Jianping Cai, Guoxin Hu
ABSTRACT
Cytochrome P450 2D6 (CYP2D6), a member of the CYP450 enzyme super family, is a polymorphic enzyme that metabolizes ~25% of therapeutic drugs. CYP2D6 exhibits significant genetic polymorphisms which might cause adverse effects and therapeutic failures of some drugs. The purpose of this study was to evaluate the catalytic activities of 22 novel CYP2D6 alleles (CYP2D6*87, *88, *89, *90, *91, *92, *93, *94, *95, *96, *97, *98, R25Q, F164L, E215K, F219S, V327M, D336N, V342M, R344Q, R440C, R497C) on the metabolism of gefitinib in vitro. CYP2D6 variants were incubated with 1-100 μM gefitinib for 60 min at 37°C and the reaction was terminated by cooling to -80°C immediately. Gefitinib and its metabolite O-desmethyl gefitinib were analyzed by an ultra-performance liquid chromatography-tandem mass spectrometry system. Compared to CYP2D6.1, most CYP2D6 variants exhibited significantly decreased relative clearance values (from 3.11% to 79.35%), whereas CYP2D6.92 and CYP2D6.96 displayed no detectable enzyme activity. Only CYP2D6.94 exhibited a markedly increased intrinsic clearance value, and eight variants (CYP2D6.88, CYP2D6.89, CYP2D6.91, CYP2D6.97, V342M, R344Q, F219S, and F164L) showed no significant difference. In addition, 23 CYP2D6 allelic isoforms exhibited substrate inhibition trend toward gefitinib. As the first study of all the aforementioned alleles for gefitinib metabolism, these comprehensive data may help in the clinical assessment of the metabolism of gefitinib, and may also offer a reference for personalized treatment with gefitinib in clinical settings.
Sigma-Aldrich
O-Desmethyl gefitinib, ≥95%, research grade