Merck
  • Hepatic NADH reductive stress underlies common variation in metabolic traits.
Hepatic NADH reductive stress underlies common variation in metabolic traits.
Nature (2020-05-29)
Russell P Goodman, Andrew L Markhard, Hardik Shah, Rohit Sharma, Owen S Skinner, Clary B Clish, Amy Deik, Anupam Patgiri, Yu-Han H Hsu, Ricard Masia, Hye Lim Noh, Sujin Suk, Olga Goldberger, Joel N Hirschhorn, Gary Yellen, Jason K Kim, Vamsi K Mootha
ABSTRACT
The cellular NADH/NAD+ ratio is fundamental to biochemistry, but the extent to which it reflects versus drives metabolic physiology in vivo is poorly understood. Here we report the in vivo application of Lactobacillus brevis (Lb)NOX1, a bacterial water-forming NADH oxidase, to assess the metabolic consequences of directly lowering the hepatic cytosolic NADH/NAD+ ratio in mice. By combining this genetic tool with metabolomics, we identify circulating α-hydroxybutyrate levels as a robust marker of an elevated hepatic cytosolic NADH/NAD+ ratio, also known as reductive stress. In humans, elevations in circulating α-hydroxybutyrate levels have previously been associated with impaired glucose tolerance2, insulin resistance3 and mitochondrial disease4, and are associated with a common genetic variant in GCKR5, which has previously been associated with many seemingly disparate metabolic traits. Using LbNOX, we demonstrate that NADH reductive stress mediates the effects of GCKR variation on many metabolic traits, including circulating triglyceride levels, glucose tolerance and FGF21 levels. Our work identifies an elevated hepatic NADH/NAD+ ratio as a latent metabolic parameter that is shaped by human genetic variation and contributes causally to key metabolic traits and diseases. Moreover, it underscores the utility of genetic tools such as LbNOX to empower studies of 'causal metabolism'.
Product Description
Avanti
12:0 PC (DLPC), Avanti Research - A Croda Brand
Avanti
12:0 PC (DLPC), 1,2-dilauroyl-sn-glycero-3-phosphocholine, chloroform
Sigma-Aldrich
BCG Albumin Assay Kit, sufficient for 250 colorimetric tests