Merck
  • Characterization of an amino acid based biodegradable surfactant facilitating the incorporation of DNA into lipophilic delivery systems.
Characterization of an amino acid based biodegradable surfactant facilitating the incorporation of DNA into lipophilic delivery systems.
Journal of colloid and interface science (2020-02-02)
Julian Dominik Wolf, Markus Kurpiers, Randi Angela Baus, Roman Xaver Götz, Janine Griesser, Barbara Matuszczak, Andreas Bernkop-Schnürch
ABSTRACT
Lysine based cationic surfactants are well-tolerated tools for hydrophobic ion pairing (HIP) with DNA and its incorporation into lipophilic delivery systems. Di-Boc-lysine was esterified with 1-hexadecanol and the Boc-residues were cleaved off resulting in hexadecyl lysinate (HL). Subsequently, its Log POctanol/water and the critical micelle concentration (CMC) were determined. Degradability was evaluated utilizing trypsin and pancreas lipase as well as Caco-2 cells. Afterwards, the viability of Caco-2 cells upon incubation with HL was investigated. Finally, HL was ion-paired with plasmid DNA (pDNA, 6159 bp) and the obtained complex was incorporated into self-emulsifying drug delivery systems (SEDDS) for transfection studies on HEK-293 cells. HL was synthesized with a yield of 53% and subsequent characterization revealed a Log PWater/Octanol of 0.05 and a CMC of 2.7 mM. Enzymatic degradation studies showed rapid degradation of HL by isolated enzymes and Caco-2 cells and cell viability experiments revealed no toxic effect of HL even in a concentration of 250 µg·ml-1 within 24 h. HIP with pDNA was the most efficient in a molar ratio of 6159:1 (HL:pDNA) equalling a charge ratio of 1:1. Formed complexes could be incorporated into SEDDS facilitating successful transfection of HEK-293 cells.
Sigma-Aldrich
N,N′-Dicyclohexylurea, 98%