Merck
  • miR-4651 inhibits cell proliferation of gingival mesenchymal stem cells by inhibiting HMGA2 under nifedipine treatment.
miR-4651 inhibits cell proliferation of gingival mesenchymal stem cells by inhibiting HMGA2 under nifedipine treatment.
International journal of oral science (2020-04-02)
Xiao Han, Ruzhuang Yang, Haoqing Yang, Yangyang Cao, Nannan Han, Chen Zhang, Ruitang Shi, Zhengting Zhang, Zhipeng Fan
ABSTRACT
Drug-induced gingival overgrowth (DIGO) is recognized as a side effect of nifedipine (NIF); however, the underlying molecular mechanisms remain unknown. In this study, we found that overexpressed miR-4651 inhibits cell proliferation and induces G0/G1-phase arrest in gingival mesenchymal stem cells (GMSCs) with or without NIF treatment. Furthermore, sequential window acquisition of all theoretical mass spectra (SWATH-MS) analysis, bioinformatics analysis, and dual-luciferase report assay results confirmed that high-mobility group AT-hook 2 (HMGA2) is the downstream target gene of miR-4651. Overexpression of HMGA2 enhanced GMSC proliferation and accelerated the cell cycle with or without NIF treatment. The present study demonstrates that miR-4651 inhibits the proliferation of GMSCs and arrests the cell cycle at the G0/G1 phase by upregulating cyclin D and CDK2 while downregulating cyclin E through inhibition of HMGA2 under NIF stimulation. These findings reveal a novel mechanism regulating DIGO progression and suggest the potential of miR-4651 and HMGA2 as therapeutic targets.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Anti-PLCγ-1 Antibody, Upstate®, from mouse
Sigma-Aldrich
Anti-Cdk4 Antibody, clone DCS-35, clone DCS-35, Chemicon®, from mouse
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Cyclin E Antibody, clone HE12, clone HE12, Upstate®, from mouse