Merck
  • Diverse effects of interferon alpha on the establishment and reversal of HIV latency.
Diverse effects of interferon alpha on the establishment and reversal of HIV latency.
PLoS pathogens (2020-02-29)
Renée M Van der Sluis, Jennifer M Zerbato, Jake W Rhodes, Rachel D Pascoe, Ajantha Solomon, Nitasha A Kumar, Ashanti I Dantanarayana, Surekha Tennakoon, Jérémy Dufloo, James McMahon, Judy J Chang, Vanessa A Evans, Paul J Hertzog, Martin R Jakobsen, Andrew N Harman, Sharon R Lewin, Paul U Cameron
ABSTRACT
HIV latency is the major barrier to a cure for people living with HIV (PLWH) on antiretroviral therapy (ART) because the virus persists in long-lived non-proliferating and proliferating latently infected CD4+ T cells. Latently infected CD4+ T cells do not express viral proteins and are therefore not visible to immune mediated clearance. Therefore, identifying interventions that can reverse latency and also enhance immune mediated clearance is of high interest. Interferons (IFNs) have multiple immune enhancing effects and can inhibit HIV replication in activated CD4+ T cells. However, the effects of IFNs on the establishment and reversal of HIV latency is not understood. Using an in vitro model of latency, we demonstrated that plasmacytoid dendritic cells (pDC) inhibit the establishment of HIV latency through secretion of type I IFNα, IFNβ and IFNω but not IFNε or type III IFNλ1 and IFNλ3. However, once latency was established, IFNα but no other IFNs were able to efficiently reverse latency in both an in vitro model of latency and CD4+ T cells collected from PLWH on suppressive ART. Binding of IFNα to its receptor expressed on primary CD4+ T cells did not induce activation of the canonical or non-canonical NFκB pathway but did induce phosphorylation of STAT1, 3 and 5 proteins. STAT5 has been previously demonstrated to bind to the HIV long terminal repeat and activate HIV transcription. We demonstrate diverse effects of interferons on HIV latency with type I IFNα; inhibiting the establishment of latency but also reversing HIV latency once latency is established.
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from herring testes, Type XIV
Sigma-Aldrich
Sample Buffer, Laemmli 2× Concentrate
Sigma-Aldrich
Fluorescein (free acid), Dye content 95 %
Sigma-Aldrich
6-Phosphonohexanoic acid, 97%
Roche
cOmplete ULTRA Tablets, Mini, EDTA-free, EASYpack Protease Inhibitor Cocktail, Tablets supplied in foil blister packs.
Sigma-Aldrich
Romidepsin, ≥98% (HPLC)