Merck
  • Causal Link between n-3 Polyunsaturated Fatty Acid Deficiency and Motivation Deficits.
Causal Link between n-3 Polyunsaturated Fatty Acid Deficiency and Motivation Deficits.
Cell metabolism (2020-03-07)
Fabien Ducrocq, Roman Walle, Andrea Contini, Asma Oummadi, Baptiste Caraballo, Suzanne van der Veldt, Marie-Lou Boyer, Frank Aby, Tarson Tolentino-Cortez, Jean-Christophe Helbling, Lucy Martine, Stéphane Grégoire, Stéphanie Cabaret, Sylvie Vancassel, Sophie Layé, Jing Xuan Kang, Xavier Fioramonti, Olivier Berdeaux, Gabriel Barreda-Gómez, Elodie Masson, Guillaume Ferreira, David W L Ma, Clementine Bosch-Bouju, Véronique De Smedt-Peyrusse, Pierre Trifilieff
ABSTRACT
Reward-processing impairment is a common symptomatic dimension of several psychiatric disorders. However, whether the underlying pathological mechanisms are common is unknown. Herein, we asked if the decrease in the n-3 polyunsaturated fatty acid (PUFA) lipid species, consistently described in these pathologies, could underlie reward-processing deficits. We show that reduced n-3 PUFA biostatus in mice leads to selective motivational impairments. Electrophysiological recordings revealed increased collateral inhibition of dopamine D2 receptor-expressing medium spiny neurons (D2-MSNs) onto dopamine D1 receptor-expressing MSNs in the nucleus accumbens, a main brain region for the modulation of motivation. Strikingly, transgenically preventing n-3 PUFA deficiency selectively in D2-expressing neurons normalizes MSN collateral inhibition and enhances motivation. These results constitute the first demonstration of a causal link between a behavioral deficit and n-3 PUFA decrease in a discrete neuronal population and suggest that lower n-3 PUFA biostatus in psychopathologies could participate in the etiology of reward-related symptoms.
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Millipore
FluorSave Reagent
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O