Merck
  • An expanded library of orthogonal split inteins enables modular multi-peptide assemblies.
An expanded library of orthogonal split inteins enables modular multi-peptide assemblies.
Nature communications (2020-04-07)
Filipe Pinto, Ella Lucille Thornton, Baojun Wang
ABSTRACT
Inteins are protein segments capable of joining adjacent residues via a peptide bond. In this process known as protein splicing, the intein itself is not present in the final sequence, thus achieving scarless peptide ligation. Here, we assess the splicing activity of 34 inteins (both uncharacterized and known) using a rapid split fluorescent reporter characterization platform, and establish a library of 15 mutually orthogonal split inteins for in vivo applications, 10 of which can be simultaneously used in vitro. We show that orthogonal split inteins can be coupled to multiple split transcription factors to implement complex logic circuits in living organisms, and that they can also be used for the in vitro seamless assembly of large repetitive proteins with biotechnological relevance. Our work demonstrates the versatility and vast potential of an expanded library of orthogonal split inteins for their use in the fields of synthetic biology and protein engineering.
Sigma-Aldrich
L-(+)-Arabinose, ≥99% (GC)
Sigma-Aldrich
Glass beads, acid-washed, 425-600 μm (30-40 U.S. sieve)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, Animal Component-free
Sigma-Aldrich
Tetracycline hydrochloride, powder
Sigma-Aldrich
Ampicillin sodium salt
Sigma-Aldrich
N-(β-Ketocaproyl)-L-homoserine lactone, ≥98%
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG
Sigma-Aldrich
Breathe-Easy® sealing membrane, polyurethane membrane with acrylic adhesive pre-cut to fit standard multiwell plates.
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile