Merck
  • Lung endothelial cells are sensitive to epsilon toxin from Clostridium perfringens.
Lung endothelial cells are sensitive to epsilon toxin from Clostridium perfringens.
Veterinary research (2020-02-26)
Jonatan Dorca-Arévalo, Eduard Dorca, Benjamín Torrejón-Escribano, Marta Blanch, Mireia Martín-Satué, Juan Blasi
ABSTRACT
The pore-forming protein epsilon toxin (Etx) from Clostridium perfringens produces acute perivascular edema affecting several organs, especially the brain and lungs. Despite the toxin evident effect on microvasculature and endothelial cells, the underlying molecular and cellular mechanisms remain obscure. Moreover, no Etx-sensitive endothelial cell model has been identified to date. Here, we characterize the mouse lung endothelial cell line 1G11 as an Etx-sensitive cell line and compare it with the well-characterized Etx-sensitive Madin-Darby canine kidney epithelial cell line. Several experimental approaches, including morphological and cytotoxic assays, clearly demonstrate that the 1G11 cell line is highly sensitive to Etx and show the specific binding, oligomerization, and pore-forming activity of the toxin in these cells. Recently, the myelin and lymphocyte (MAL) protein has been postulated as a putative receptor for Etx. Here, we show the presence of Mal mRNA in the 1G11 cell line and the presence of the MAL protein in the endothelium of some mouse lung vessels, supporting the hypothesis that this protein is a key element in the Etx intoxication pathway. The existence of an Etx-sensitive cell line of endothelial origin would help shed light on the cellular and molecular mechanisms underlying Etx-induced edema and its consequences.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, sodium pyruvate, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Fluoromount Aqueous Mounting Medium, for use with fluorescent dye-stained tissues
Sigma-Aldrich
MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile