Merck
  • Characterization of novel conformation-selective α-synuclein antibodies as potential immunotherapeutic agents for Parkinson's disease.
Characterization of novel conformation-selective α-synuclein antibodies as potential immunotherapeutic agents for Parkinson's disease.
Neurobiology of disease (2019-12-15)
Michael X Henderson, Dustin J Covell, Charlotte Hiu-Yan Chung, Rose M Pitkin, Raizel M Sandler, Samantha C Decker, Dawn M Riddle, Bin Zhang, Ronald J Gathagan, Michael J James, John Q Trojanowski, Kurt R Brunden, Virginia M Y Lee, Kelvin C Luk
ABSTRACT
Parkinson's disease (PD) and dementia with Lewy bodies (DLB) are progressive neurodegenerative diseases for which there is no disease-modifying treatment. PD and DLB are characterized by aggregation of the synaptic protein α-synuclein, and there is compelling evidence to suggest that progression of these diseases is associated with the trans-cellular spread of pathogenic α-synuclein through the brains of afflicted individuals. Therapies targeting extracellular, pathogenic α-synuclein may therefore hold promise for slowing or halting disease progression. In this regard, it has been suggested that highly-selective antibodies can be administered as therapeutic agents targeting pathogenic proteins. In the current study, we screened a series of antibodies using multiple selection criterion to identify those that selectively bind pathogenic α-synuclein and show potent inhibition of pathology seeding in a neuronal model of α-synucleinopathy. A lead antibody was tested in a mouse model of PD, and it was able to reduce the spread of α-synuclein pathology in the brain and attenuate dopamine reductions in the striatum. This study highlights the therapeutic potential of α-synuclein immunotherapy for the treatment of PD and DLB, and provides a framework for screening of α-synuclein antibodies to identify those with preferred properties.
Sigma-Aldrich
Dopamine hydrochloride
Sigma-Aldrich
Azaserine-Hypoxanthine 50x, Hybri-Max, γ-irradiated, lyophilized powder, BioXtra, suitable for hybridoma
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Ammonium hydroxide solution, puriss., meets analytical specification of Ph. Eur., 25-30% NH3 basis