Merck
  • EHD1 and RUSC2 Control Basal Epidermal Growth Factor Receptor Cell Surface Expression and Recycling.
EHD1 and RUSC2 Control Basal Epidermal Growth Factor Receptor Cell Surface Expression and Recycling.
Molecular and cellular biology (2020-01-15)
Eric C Tom, Insha Mushtaq, Bhopal C Mohapatra, Haitao Luan, Aaqib M Bhat, Neha Zutshi, Sukanya Chakraborty, Namista Islam, Priyanka Arya, Timothy A Bielecki, Fany M Iseka, Sohinee Bhattacharyya, Luke R Cypher, Benjamin T Goetz, Simarjeet K Negi, Matthew D Storck, Sandeep Rana, Angelika Barnekow, Pankaj K Singh, Guoguang Ying, Chittibabu Guda, Amarnath Natarajan, Vimla Band, Hamid Band
ABSTRACT
Epidermal growth factor receptor (EGFR) is a prototype receptor tyrosine kinase and an oncoprotein in many solid tumors. Cell surface display of EGFR is essential for cellular responses to its ligands. While postactivation endocytic trafficking of EGFR has been well elucidated, little is known about mechanisms of basal/preactivation surface display of EGFR. Here, we identify a novel role of the endocytic regulator EHD1 and a potential EHD1 partner, RUSC2, in cell surface display of EGFR. EHD1 and RUSC2 colocalize with EGFR in vesicular/tubular structures and at the Golgi compartment. Inducible EHD1 knockdown reduced the cell surface EGFR expression with accumulation at the Golgi compartment, a phenotype rescued by exogenous EHD1. RUSC2 knockdown phenocopied the EHD1 depletion effects. EHD1 or RUSC2 depletion impaired the EGF-induced cell proliferation, demonstrating that the novel, EHD1- and RUSC2-dependent transport of unstimulated EGFR from the Golgi compartment to the cell surface that we describe is functionally important, with implications for physiologic and oncogenic roles of EGFR and targeted cancer therapies.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Guanosine 5′-[γ-thio]triphosphate tetralithium salt, ≥90% (contains < 10% GDP, HPLC), powder
Millipore
Immobilon®-P PVDF Membrane, 1 roll, 27 cm x 3.75 m, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.
Sigma-Aldrich
MISSION® esiRNA, targeting human RUSC2
Sigma-Aldrich
MISSION® esiRNA, targeting human EHD1 (2)