Merck
  • Alcohol dependence potentiates substance P/neurokinin-1 receptor signaling in the rat central nucleus of amygdala.
Alcohol dependence potentiates substance P/neurokinin-1 receptor signaling in the rat central nucleus of amygdala.
Science advances (2020-03-25)
S Khom, T Steinkellner, T S Hnasko, M Roberto
ABSTRACT
Behavioral and clinical studies suggest a critical role of substance P (SP)/neurokinin-1 receptor (NK-1R) signaling in alcohol dependence. Here, we examined regulation of GABA transmission in the medial subdivision of the central amygdala (CeM) by the SP/NK-1R system, and its neuroadaptation following chronic alcohol exposure. In naïve rats, SP increased action potential-dependent GABA release, and the selective NK-1R antagonist L822429 decreased it, demonstrating SP regulation of CeM activity under basal conditions. SP induced a larger GABA release in alcohol-dependent rats accompanied by decreased NK-1R expression compared to naïve controls, suggesting NK-1R hypersensitivity which persisted during protracted alcohol withdrawal. The NK-1R antagonist blocked acute alcohol-induced GABA release in alcohol-dependent and withdrawn but not in naïve rats, indicating that dependence engages the SP/NK-1R system to mediate acute effects of alcohol. Collectively, we report long-lasting CeA NK-1R hypersensitivity corroborating that NK-1Rs are promising targets for the treatment of alcohol use disorder.
Sigma-Aldrich
Bisindolylmaleimide XI hydrochloride, ≥98% (TLC), solid
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
DNQX, ≥98% (TLC)