Merck
gamma-Cyclodextrin: a review on enzymatic production and applications.
Applied microbiology and biotechnology (2007-09-25)
Zhaofeng Li, Miao Wang, Feng Wang, Zhengbiao Gu, Guocheng Du, Jing Wu, Jian Chen
ABSTRACT
Cyclodextrins are cyclic alpha-1,4-glucans that are produced from starch or starch derivates using cyclodextrin glycosyltransferase (CGTase). The most common forms are alpha-, beta-, and gamma-cyclodextrins. This mini-review focuses on the enzymatic production, unique properties, and applications of gamma-cyclodextrin as well as its difference with alpha- and beta-cyclodextrins. As all known wild-type CGTases produce a mixture of alpha-, beta-, and gamma-cyclodextrins, the obtaining of a CGTase predominantly producing gamma-cyclodextrin is discussed. Recently, more economic production processes for gamma-cyclodextrin have been developed using improved gamma-CGTases and appropriate complexing agents. Compared with alpha- and beta-cyclodextrins, gamma-cyclodextrin has a larger internal cavity, higher water solubility, and more bioavailability, so it has wider applications in many industries, especially in the food and pharmaceutical industries.
Sigma-Aldrich
γ-Cyclodextrin, Produced by Wacker Chemie AG, Burghausen, Germany, Life Science, 98.3-102.0% cyclodextrin basis
Sigma-Aldrich
β-Cyclodextrin, ≥97%
Sigma-Aldrich
γ-Cyclodextrin, powder, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
γ-Cyclodextrin, ≥98%
Sigma-Aldrich
γ-Cyclodextrin, produced by Wacker Chemie AG, Burghausen, Germany, ≥90.0% cyclodextrin basis (HPLC)
Gammacyclodextrin, European Pharmacopoeia (EP) Reference Standard