Merck
  • A self-aggregating peptide: implications for the development of thermostable vaccine candidates.
A self-aggregating peptide: implications for the development of thermostable vaccine candidates.
BMC biotechnology (2020-01-22)
Adolfo Cruz-Reséndiz, Jesús Zepeda-Cervantes, Alicia Sampieri, Carlos Bastián-Eugenio, Gonzalo Acero, J Iván Sánchez-Betancourt, Goar Gevorkian, Luis Vaca
ABSTRACT
The use of biomaterials has been expanded to improve the characteristics of vaccines. Recently we have identified that the peptide PH(1-110) from polyhedrin self-aggregates and incorporates foreign proteins to form particles. We have proposed that this peptide can be used as an antigen carrying system for vaccines. However, the immune response generated by the antigen fused to the peptide has not been fully characterized. In addition, the adjuvant effect and thermostability of the particles has not been evaluated. In the present study we demonstrate the use of a system developed to generate nano and microparticles carrying as a fusion protein peptides or proteins of interest to be used as vaccines. These particles are purified easily by centrifugation. Immunization of animals with the particles in the absence of adjuvant result in a robust and long-lasting immune response. Proteins contained inside the particles are maintained for over 1 year at ambient temperature, preserving their immunological properties. The rapid and efficient production of the particles in addition to the robust immune response they generate position this system as an excellent method for the rapid response against emerging diseases. The thermostability conferred by the particle system facilitates the distribution of the vaccines in developing countries or areas with no electricity.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Freund′s Adjuvant, Incomplete, liquid
Sigma-Aldrich
Freund′s Adjuvant, Complete, cell suspension
Sigma-Aldrich
Nitrilotriacetic acid disodium salt, Sigma Grade, ≥99%
Millipore
Immobilon®-NC Transfer Membrane, 1 roll, 33 cm x 3 m, 0.45 µm pore size, Triton-free, mixed cellulose esters transfer membrane
Millipore
Green Fluorescent Protein