Merck
Functionalized lipids and surfactants for specific applications.
Biochimica et biophysica acta (2016-03-08)
Mariusz Kepczynski, Tomasz Róg
ABSTRACT
Synthetic lipids and surfactants that do not exist in biological systems have been used for the last few decades in both basic and applied science. The most notable applications for synthetic lipids and surfactants are drug delivery, gene transfection, as reporting molecules, and as support for structural lipid biology. In this review, we describe the potential of the synergistic combination of computational and experimental methodologies to study the behavior of synthetic lipids and surfactants embedded in lipid membranes and liposomes. We focused on select cases in which molecular dynamics simulations were used to complement experimental studies aiming to understand the structure and properties of new compounds at the atomistic level. We also describe cases in which molecular dynamics simulations were used to design new synthetic lipids and surfactants, as well as emerging fields for the application of these compounds. This article is part of a Special Issue entitled: Biosimulations edited by Ilpo Vattulainen and Tomasz Róg.
Product Description
Avanti
16:0 Glutaryl PE, Avanti Research - A Croda Brand 870245P, powder
Avanti
16:0 Biotinyl PE, Avanti Research - A Croda Brand 870285P, powder
Avanti
18:1 Succinyl PE, Avanti Research - A Croda Brand 870222P, powder
Avanti
18:1 PDP PE, Avanti Research - A Croda Brand 870202O
Avanti
16:0 Succinyl PE, Avanti Research - A Croda Brand 870225P, powder
Avanti
18:1 Caproylamine PE, Avanti Research - A Croda Brand
Avanti
16:0 PDP PE, Avanti Research - A Croda Brand 870205P, powder