Merck
  • Ethanolamine and Phosphatidylethanolamine: Partners in Health and Disease.
Ethanolamine and Phosphatidylethanolamine: Partners in Health and Disease.
Oxidative medicine and cellular longevity (2017-08-09)
Dhaval Patel, Stephan N Witt
ABSTRACT
Phosphatidylethanolamine (PE) is the second most abundant phospholipid in mammalian cells. PE comprises about 15-25% of the total lipid in mammalian cells; it is enriched in the inner leaflet of membranes, and it is especially abundant in the inner mitochondrial membrane. PE has quite remarkable activities: it is a lipid chaperone that assists in the folding of certain membrane proteins, it is required for the activity of several of the respiratory complexes, and it plays a key role in the initiation of autophagy. In this review, we focus on PE's roles in lipid-induced stress in the endoplasmic reticulum (ER), Parkinson's disease (PD), ferroptosis, and cancer.
Product Description
Avanti
08:0 PE, 1,2-dioctanoyl-sn-glycero-3-phosphoethanolamine, powder
Avanti
16:1 PE, 1,2-dipalmitoleoyl-sn-glycero-3-phosphoethanolamine, chloroform
Avanti
06:0 PE, 1,2-dihexanoyl-sn-glycero-3-phosphoethanolamine, powder
Avanti
14:0 PE, Avanti Research - A Croda Brand
Avanti
TopFluor TMR PE, Avanti Research - A Croda Brand 810241P, powder
Avanti
18:0-18:1 PE, 1-stearoyl-2-oleoyl-sn-glycero-3-phosphoethanolamine, powder
Avanti
15:0-18:1-d7-PE, Avanti Research - A Croda Brand