Merck
  • Antiproliferative Effects of Thymoquinone in MCF-7 Breast and HepG2 Liver Cancer Cells: Possible Role of Ceramide and ER Stress.
Antiproliferative Effects of Thymoquinone in MCF-7 Breast and HepG2 Liver Cancer Cells: Possible Role of Ceramide and ER Stress.
Nutrition and cancer (2020-04-15)
Mutay Aslan, Ebru Afşar, Esma Kırımlıoglu, Tuğçe Çeker, Çağatay Yılmaz
ABSTRACT
We aimed to investigate the impact of thymoquinone (TQ), on sphingolipid metabolites, ER stress and apoptotic pathways in MCF-7 and HepG2 cancer cells. Antiproliferative effect was exerted in cancer cells via TQ incubation at different doses and durations. Cell viability was measured by MTT assay. Levels of sphingosine-1-phosphate (S1P), C16-C24 sphingomyelins (SM) and C16-C24 ceramides (CER) were determined by LC-MS/MS. Neutral sphingomyelinase (N-SMase) enzyme activity was measured by colorimetric assay and ceramide-1-phosphate (C1P) levels were determined by immunoassay. Nuclear factor kappa-b subunit 1 (NFκB1) and glucose-regulated protein 78-kd (GRP78) gene expressions were evaluated by quantitative PCR analysis, while NF-κB p65, GRP 78 and cleaved caspase-3 protein levels were assesed by immunofluorescence and western blot analysis. Incubation with TQ significantly decreased cell viability, S1P, C1P, NF-κB1 mRNA and NF-κB p65 protein levels in cancer cells compared to controls. A significant increase was observed in N-SMase activity, cellular levels of C16-C24 CERs and cleaved caspase-3 levels in cancer cells treated with TQ. GRP78 mRNA and protein levels also increased in cancer cells treated with TQ. In conclusion, TQ-induced ceramide accumulation and ER stress in conjunction with decreased S1P, C1P and NF-κB mediated cell survival may promote cancer cell death by triggering apoptosis.
Product Description
Avanti
C18 Ceramide (d18:1/18:0), Avanti Research - A Croda Brand
Avanti
C20 Ceramide (d18:1/20:0), Avanti Research - A Croda Brand
Avanti
C24 Ceramide (d18:1/24:0), Avanti Research - A Croda Brand
Avanti
C16 Ceramide (d14:1/16:0), Avanti Research - A Croda Brand, powder
Avanti
C22 Ceramide (d18:1/22:0), Avanti Research - A Croda Brand 860501P, powder