Merck
  • Native Chromatin Proteomics Reveals a Role for Specific Nucleoporins in Heterochromatin Organization and Maintenance.
Native Chromatin Proteomics Reveals a Role for Specific Nucleoporins in Heterochromatin Organization and Maintenance.
Molecular cell (2019-12-01)
Nahid Iglesias, Joao A Paulo, Antonis Tatarakis, Xiaoyi Wang, Amanda L Edwards, Natarajan V Bhanu, Benjamin A Garcia, Wilhelm Haas, Steven P Gygi, Danesh Moazed
ABSTRACT
Spatially and functionally distinct domains of heterochromatin and euchromatin play important roles in the maintenance of chromosome stability and regulation of gene expression, but a comprehensive knowledge of their composition is lacking. Here, we develop a strategy for the isolation of native Schizosaccharomyces pombe heterochromatin and euchromatin fragments and analyze their composition by using quantitative mass spectrometry. The shared and euchromatin-specific proteomes contain proteins involved in DNA and chromatin metabolism and in transcription, respectively. The heterochromatin-specific proteome includes all proteins with known roles in heterochromatin formation and, in addition, is enriched for subsets of nucleoporins and inner nuclear membrane (INM) proteins, which associate with different chromatin domains. While the INM proteins are required for the integrity of the nucleolus, containing ribosomal DNA repeats, the nucleoporins are required for aggregation of heterochromatic foci and epigenetic inheritance. The results provide a comprehensive picture of heterochromatin-associated proteins and suggest a role for specific nucleoporins in heterochromatin function.
Sigma-Aldrich
Dess-Martin periodinane, 97%
Sigma-Aldrich
Ethylene glycol-bis(succinic acid N-hydroxysuccinimide ester), powder
Sigma-Aldrich
EPPS, ≥99.5% (titration)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys14) Antibody, serum, Upstate®
Supelco
Pesticide standard 17 solution, PESTANAL®, analytical standard, according to DIN 38407, in acetonitrile
Sigma-Aldrich
Urea, powder, BioReagent, suitable for cell culture
Supelco
Empore SPE Disks, matrix active group C18, diam. 47 mm, pk of 20