Merck
  • Dissecting the Regulatory Strategies of NF-κB RelA Target Genes in the Inflammatory Response Reveals Differential Transactivation Logics.
Dissecting the Regulatory Strategies of NF-κB RelA Target Genes in the Inflammatory Response Reveals Differential Transactivation Logics.
Cell reports (2020-02-27)
Kim A Ngo, Kensei Kishimoto, Jeremy Davis-Turak, Aditya Pimplaskar, Zhang Cheng, Roberto Spreafico, Emily Y Chen, Amy Tam, Gourisankar Ghosh, Simon Mitchell, Alexander Hoffmann
ABSTRACT
Nuclear factor κB (NF-κB) RelA is the potent transcriptional activator of inflammatory response genes. We stringently defined a list of direct RelA target genes by integrating physical (chromatin immunoprecipitation sequencing [ChIP-seq]) and functional (RNA sequencing [RNA-seq] in knockouts) datasets. We then dissected each gene's regulatory strategy by testing RelA variants in a primary-cell genetic-complementation assay. All endogenous target genes require RelA to make DNA-base-specific contacts, and none are activatable by the DNA binding domain alone. However, endogenous target genes differ widely in how they employ the two transactivation domains. Through model-aided analysis of the dynamic time-course data, we reveal the gene-specific synergy and redundancy of TA1 and TA2. Given that post-translational modifications control TA1 activity and intrinsic affinity for coactivators determines TA2 activity, the differential TA logics suggests context-dependent versus context-independent control of endogenous RelA-target genes. Although some inflammatory initiators appear to require co-stimulatory TA1 activation, inflammatory resolvers are a part of the NF-κB RelA core response.
Sigma-Aldrich
Poly(deoxyinosinic-deoxycytidylic) acid sodium salt, double-stranded alternating copolymer
Sigma-Aldrich
Di(N-succinimidyl) glutarate, ≥97.0% (CHN)